ADVH303BC11F AChemBlock

2-methoxy-4,6-dimethylpyrimidin-5-ol

  • MDL number MFCD08276902
  • Cas Number : 345642-89-9
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methoxy-4-6-dimethylpyrimidin-5-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62956
Availability details for
Quantity
  • Availability
  • Add to Cart