ADVH303BC16D AChemBlock

(R)-4-(tert-butoxycarbonyl)-4-azaspiro[2.4]heptane-5-carboxylic acid

  • MDL number MFCD30470758
  • Cas Number : 2007916-06-3
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-4-tert-butoxycarbonyl-4-azaspiro-2-4-heptane-5-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63036
Availability details for
Quantity
  • Availability
  • Add to Cart