ADVH303BC19A AChemBlock

tert-butyl 8-oxa-2,5-diazaspiro[3.5]nonane-5-carboxylate

  • MDL number MFCD14581203
  • Cas Number : 1251020-41-3
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-8-oxa-2-5-diazaspiro-3-5-nonane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63082
Availability details for
Quantity
  • Availability
  • Add to Cart