ADVH303BC1D9 AChemBlock

(4-chlorothieno[3,2-d]pyrimidin-6-yl)methanol

  • MDL number MFCD12923082
  • Cas Number : 1207972-75-5
  • Molecular Weight : 200.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chlorothieno-3-2-d-pyrimidin-6-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63146
Availability details for
Quantity
  • Availability
  • Add to Cart