ADVH303BC1E8 AChemBlock

tert-butyl ((1S,4s,6S)-6-aminospiro[3.3]heptan-1-yl)carbamate

  • Cas Number : 1682624-72-1
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1s-4s-6s-6-aminospiro-3-3-heptan-1-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63162
Availability details for
Quantity
  • Availability
  • Add to Cart