ADVH303BC227 AChemBlock

tert-butyl (R)-3,3-difluoro-1,6-diazaspiro[3.4]octane-6-carboxylate

  • Cas Number : 2166191-89-3
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-r-3-3-difluoro-1-6-diazaspiro-3-4-octane-6-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63227
Availability details for
Quantity
  • Availability
  • Add to Cart