ADVH303BC2CD AChemBlock

tert-butyl (3S,4S)-3-(hydroxymethyl)-4-methylpyrrolidine-1-carboxylate

  • MDL number MFCD12407173
  • Cas Number : 1417789-75-3
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-4s-3-hydroxymethyl-4-methylpyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62523
Availability details for
Quantity
  • Availability
  • Add to Cart