ADVH303BC2E3 AChemBlock

tert-butyl (R)-3-fluoro-3-(hydroxymethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD30803203
  • Cas Number : 1893340-46-9
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-r-3-fluoro-3-hydroxymethyl-pyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62528
Availability details for
Quantity
  • Availability
  • Add to Cart