ADVH303BC3E7 AChemBlock

(2R,3S)-3-fluoropyrrolidine-2-carboxylic acid compound with 2,2,2-trifluoroacetic acid (1:1)

  • Cas Number : 926910-60-3
  • Molecular Weight : 247.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2r-3s-3-fluoropyrrolidine-2-carboxylic-acid-compound-with-2-2-2-trifluoroacetic-acid-1-1.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q64267
Availability details for
Quantity
  • Availability
  • Add to Cart