ADVH303BC4FF AChemBlock

tert-butyl (R)-3,3-difluoro-4-hydroxypiperidine-1-carboxylate

  • Cas Number : 1893408-14-4
  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-r-3-3-difluoro-4-hydroxypiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63848
Availability details for
Quantity
  • Availability
  • Add to Cart