ADVH303BC522 AChemBlock

(S)-4,4-difluoropyrrolidine-2-carbonitrile hydrochloride

  • Cas Number : 869489-04-3
  • Molecular Weight : 168.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/s-4-4-difluoropyrrolidine-2-carbonitrile-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63521
Availability details for
Quantity
  • Availability
  • Add to Cart