ADVH303BC566 AChemBlock

6-((tert-butoxycarbonyl)amino)bicyclo[3.1.0]hexane-3-carboxylic acid

  • MDL number MFCD31729125
  • Cas Number : 2168763-80-0
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-tert-butoxycarbonyl-amino-bicyclo-3-1-0-hexane-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63953
Availability details for
Quantity
  • Availability
  • Add to Cart