ADVH303BC5B9 AChemBlock

(R)-2-methyl-5-oxopyrrolidine-2-carboxylic acid

  • Cas Number : 1629052-62-5
  • Molecular Weight : 143.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-2-methyl-5-oxopyrrolidine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q64038
Availability details for
Quantity
  • Availability
  • Add to Cart