ADVH303BC5C3 AChemBlock

1,1-difluoro-8-azaspiro[4.5]decane

  • MDL number MFCD22209541
  • Cas Number : 1357354-36-9
  • Molecular Weight : 175.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-1-difluoro-8-azaspiro-4-5-decane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63469
Availability details for
Quantity
  • Availability
  • Add to Cart