ADVH303BC5EF AChemBlock

tert-butyl (3R,5S)-3-amino-5-hydroxypiperidine-1-carboxylate

  • Cas Number : 1932513-59-1
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-5s-3-amino-5-hydroxypiperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q64094
Availability details for
Quantity
  • Availability
  • Add to Cart