ADVH303BC608 AChemBlock

tert-butyl 1-amino-3-azabicyclo[3.2.0]heptane-3-carboxylate

  • MDL number MFCD14581339
  • Cas Number : 1251009-41-2
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-amino-3-azabicyclo-3-2-0-heptane-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q64120
Availability details for
Quantity
  • Availability
  • Add to Cart