ADVH303BC61A AChemBlock

(3aS,6aS)-tetrahydro-1H-cyclopenta[c]furan-5(3H)-one

  • Cas Number : 187528-23-0
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3as-6as-tetrahydro-1h-cyclopenta-c-furan-5-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q64139
Availability details for
Quantity
  • Availability
  • Add to Cart