ADVH303BC94D AChemBlock

(1R,3R)-3-amino-N-methylcyclopentane-1-carboxamide hydrochloride

  • Molecular Weight : 178.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1r-3r-3-amino-n-methylcyclopentane-1-carboxamide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K13472
Availability details for
Quantity
  • Availability
  • Add to Cart