ADVH303BCA09 AChemBlock

(S)-5-(hydroxymethyl)-1-methylpyrrolidin-2-one

  • MDL number MFCD19690658
  • Cas Number : 122663-19-8
  • Molecular Weight : 129.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/s-5-hydroxymethyl-1-methylpyrrolidin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q64629
Availability details for
Quantity
  • Availability
  • Add to Cart