ADVH303BCA5E AChemBlock

1-(tert-butyl) 4-methyl 2-methylpiperidine-1,4-dicarboxylate

  • MDL number MFCD29919080
  • Cas Number : 1415740-83-8
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-4-methyl-2-methylpiperidine-1-4-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q64987
Availability details for
Quantity
  • Availability
  • Add to Cart