ADVH303BCAAA AChemBlock

1-(tert-butyl) 3-methyl (3S,5R)-5-hydroxypiperidine-1,3-dicarboxylate

  • Cas Number : 1246442-45-4
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-3-methyl-3s-5r-5-hydroxypiperidine-1-3-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65067
Availability details for
Quantity
  • Availability
  • Add to Cart