ADVH303BCADD AChemBlock

tert-butyl 6-oxa-2,9-diazaspiro[4.5]decane-2-carboxylate

  • MDL number MFCD17017735
  • Cas Number : 1366647-28-0
  • Molecular Weight : 242.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-6-oxa-2-9-diazaspiro-4-5-decane-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65131
Availability details for
Quantity
  • Availability
  • Add to Cart