ADVH303BCBDF AChemBlock

tert-butyl (R)-3-amino-3-methylpyrrolidine-1-carboxylate

  • Cas Number : 1817789-07-3
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-r-3-amino-3-methylpyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K16113
Availability details for
Quantity
  • Availability
  • Add to Cart