ADVH303BCBEA AChemBlock

4-fluoro-2-methoxy-5-nitroaniline

  • MDL number MFCD23098941
  • Cas Number : 1075705-01-9
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-fluoro-2-methoxy-5-nitroaniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65476
Availability details for
Quantity
  • Availability
  • Add to Cart