ADVH303C20C3 AChemBlock

(S)-2-Methoxymethyl-morpholine hydrochloride

  • MDL number MFCD16294688
  • Cas Number : 141196-38-5
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form white solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/s-2-methoxymethyl-morpholine-hydrochloride.html

Safety Information

signalword warning
pcodes P261;P280;P305+351+338
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=B-1097
pictograms GHS07
hstatements H302;H335
Availability details for
Quantity
  • Availability
  • Add to Cart