ADVH303C2145 AChemBlock

(R)-1-(naphthalen-2-yl)ethan-1-amine

  • MDL number MFCD00085367
  • Cas Number : 3906-16-9
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-1-naphthalen-2-yl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q52610
Availability details for
Quantity
  • Availability
  • Add to Cart