ADVH303C21A7 AChemBlock

tert-butyl 3-amino-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate

  • MDL number MFCD07371643
  • Cas Number : 355819-02-2
  • Molecular Weight : 249.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-amino-7-8-dihydro-1-6-naphthyridine-6-5h-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q53554
Availability details for
Quantity
  • Availability
  • Add to Cart