ADVH303C226E AChemBlock

methyl 1-aminocyclopentane-1-carboxylate hydrochloride

  • MDL number MFCD03840364
  • Cas Number : 60421-23-0
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-aminocyclopentane-1-carboxylate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q56244
Availability details for
Quantity
  • Availability
  • Add to Cart