ADVH303C232A AChemBlock

2,4-dichloro-6-methyl-3-nitropyridine

  • MDL number MFCD00218548
  • Cas Number : 63897-12-1
  • Molecular Weight : 207.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-6-methyl-3-nitropyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q62576
Availability details for
Quantity
  • Availability
  • Add to Cart