ADVH303C2347 AChemBlock

2,2-dimethyldihydrofuran-3(2H)-one

  • MDL number MFCD09834515
  • Cas Number : 52662-40-5
  • Molecular Weight : 114.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-2-dimethyldihydrofuran-3-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q63498
Availability details for
Quantity
  • Availability
  • Add to Cart