ADVH303C246B AChemBlock

methyl 1-methyl-2-nitro-1H-imidazole-5-carboxylate

  • MDL number MFCD06738745
  • Cas Number : 40361-79-3
  • Molecular Weight : 185.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-methyl-2-nitro-1h-imidazole-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65569
Availability details for
Quantity
  • Availability
  • Add to Cart