ADVH303C246F AChemBlock

methyl 3-(2-(1,3-dioxoisoindolin-2-yl)ethyl)-6-fluoro-1H-indole-4-carboxylate

  • MDL number MFCD30478789
  • Cas Number : 1408282-25-6
  • Molecular Weight : 366.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-2-1-3-dioxoisoindolin-2-yl-ethyl-6-fluoro-1h-indole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65573
Availability details for
Quantity
  • Availability
  • Add to Cart