ADVH303C2474 AChemBlock

tert-butyl (1-(2-(5-((3-methyloxetan-3-yl)methoxy)-1H-benzo[d]imidazol-1-yl)quinolin-8-yl)piperidin-4-yl)carbamate

  • MDL number MFCD31699917
  • Cas Number : 816463-40-8
  • Molecular Weight : 543.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-2-5-3-methyloxetan-3-yl-methoxy-1h-benzo-d-imidazol-1-yl-quinolin-8-yl-piperidin-4-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65578
Availability details for
Quantity
  • Availability
  • Add to Cart