ADVH303C2476 AChemBlock

2,3,4,5-tetrahydro-1H-1,5-methanobenzo[d]azepine hydrochloride

  • MDL number MFCD20278229
  • Cas Number : 230615-52-8
  • Molecular Weight : 195.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/2-3-4-5-tetrahydro-1h-1-5-methanobenzo-d-azepine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65580
Availability details for
Quantity
  • Availability
  • Add to Cart