ADVH303C2479 AChemBlock

4-phenyl-1H-pyrazole-3-carbonitrile

  • MDL number MFCD20486377
  • Cas Number : 21673-04-1
  • Molecular Weight : 169.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/4-phenyl-1h-pyrazole-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65583
Availability details for
Quantity
  • Availability
  • Add to Cart