ADVH303C247C AChemBlock

tert-butyl thiophen-2-ylcarbamate

  • MDL number MFCD02677742
  • Cas Number : 56267-50-6
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-thiophen-2-ylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65586
Availability details for
Quantity
  • Availability
  • Add to Cart