ADVH303C2480 AChemBlock

2-(2-bromophenyl)-1H-imidazole

  • MDL number MFCD08668745
  • Cas Number : 162356-38-9
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-bromophenyl-1h-imidazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65590
Availability details for
Quantity
  • Availability
  • Add to Cart