ADVH303C2486 AChemBlock

methyl 2-cyano-4-methylbenzoate

  • MDL number MFCD18399107
  • Cas Number : 1213767-07-7
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-cyano-4-methylbenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65596
Availability details for
Quantity
  • Availability
  • Add to Cart