ADVH303C2490 AChemBlock

5-chloro-2,4-dihydroxybenzaldehyde

  • MDL number MFCD18324145
  • Cas Number : 131088-02-3
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-2-4-dihydroxybenzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65606
Availability details for
Quantity
  • Availability
  • Add to Cart