ADVH303C2501 AChemBlock

(1H-benzo[d][1,2,3]triazol-1-yl)(2,4-dichlorophenyl)methanone

  • MDL number MFCD00195103
  • Cas Number : 200626-61-5
  • Molecular Weight : 292.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/1h-benzo-d-1-2-3-triazol-1-yl-2-4-dichlorophenyl-methanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65719
Availability details for
Quantity
  • Availability
  • Add to Cart