ADVH303C2503 AChemBlock

sodium 2-((4-(3-methoxyphenyl)-5-(pyridin-2-yl)-4H-1,2,4-triazol-3-yl)thio)acetate

  • Cas Number : 1459687-89-8
  • Molecular Weight : 364.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/sodium-2-4-3-methoxyphenyl-5-pyridin-2-yl-4h-1-2-4-triazol-3-yl-thio-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65721
Availability details for
Quantity
  • Availability
  • Add to Cart