ADVH303C254A AChemBlock

(E)-4-methoxy-N-(2-(2-(pyridin-4-yl)vinyl)phenyl)benzenesulfonamide

  • MDL number MFCD00945104
  • Cas Number : 173528-92-2
  • Molecular Weight : 366.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/e-4-methoxy-n-2-2-pyridin-4-yl-vinyl-phenyl-benzenesulfonamide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65792
Availability details for
Quantity
  • Availability
  • Add to Cart