ADVH303C2557 AChemBlock

N-((1-methylpiperidin-4-yl)methyl)-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazin-6-amine

  • MDL number MFCD16659064
  • Cas Number : 1025065-69-3
  • Molecular Weight : 405.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/n-1-methylpiperidin-4-yl-methyl-3-3-trifluoromethoxy-phenyl-imidazo-1-2-b-pyridazin-6-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q65806
Availability details for
Quantity
  • Availability
  • Add to Cart