ADVH303C261F AChemBlock

2-(pyrrolidine-1-carbonyl)benzoic acid

  • MDL number MFCD01961578
  • Cas Number : 20320-43-8
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-pyrrolidine-1-carbonyl-benzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q66013
Availability details for
Quantity
  • Availability
  • Add to Cart