ADVH303C269C AChemBlock

methyl 2,5-dioxo-2,5-dihydro-1H-pyrrole-1-carboxylate

  • MDL number MFCD00042755
  • Cas Number : 55750-48-6
  • Molecular Weight : 155.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-5-dioxo-2-5-dihydro-1h-pyrrole-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q66138
Availability details for
Quantity
  • Availability
  • Add to Cart