ADVH303C3147 AChemBlock

2-(5-((3-methyloxetan-3-yl)methoxy)-1H-benzo[d]imidazol-1-yl)quinolin-8-ol

  • MDL number MFCD31699916
  • Cas Number : 816463-38-4
  • Molecular Weight : 361.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-5-3-methyloxetan-3-yl-methoxy-1h-benzo-d-imidazol-1-yl-quinolin-8-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q68873
Availability details for
Quantity
  • Availability
  • Add to Cart