ADVH303C3148 AChemBlock

2-bromo-6-methyl-3-nitropyridine

  • MDL number MFCD03095212
  • Cas Number : 374633-31-5
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-6-methyl-3-nitropyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q68874
Availability details for
Quantity
  • Availability
  • Add to Cart