ADVH303C3149 AChemBlock

3,5-dichloro-1-methylpyrazin-2(1H)-one

  • MDL number MFCD12022618
  • Cas Number : 87486-33-7
  • Molecular Weight : 179.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/3-5-dichloro-1-methylpyrazin-2-1h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q68875
Availability details for
Quantity
  • Availability
  • Add to Cart