ADVH303C3252 AChemBlock

N-((benzyloxy)carbonyl)-N-methyl-L-leucine

  • Cas Number : 33099-08-0
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/n-benzyloxy-carbonyl-n-methyl-l-leucine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q69140
Availability details for
Quantity
  • Availability
  • Add to Cart