ADVH303C32EC AChemBlock

1-(3-(((2,5-dioxopyrrolidin-1-yl)oxy)carbonyl)benzyl)-4-(5-(4-methoxyphenyl)oxazol-2-yl)pyridin-1-ium bromide

  • MDL number MFCD00467811
  • Cas Number : 155862-95-6
  • Molecular Weight : 564.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-2-5-dioxopyrrolidin-1-yl-oxy-carbonyl-benzyl-4-5-4-methoxyphenyl-oxazol-2-yl-pyridin-1-ium-bromide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=Q69294
Availability details for
Quantity
  • Availability
  • Add to Cart